  • Search Results
  • Validation of a hypoxia-inducible factor-1 alpha specimen collection procedure and quantitative enzyme-linked immunosorbent assay in solid tumor tissues.
Validation of a hypoxia-inducible factor-1 alpha specimen collection procedure and quantitative enzyme-linked immunosorbent assay in solid tumor tissues.
Analytical biochemistry (2014-05-07)
Sook Ryun Park, Robert J Kinders, Sonny Khin, Melinda Hollingshead, Smitha Antony, Ralph E Parchment, Joseph E Tomaszewski, Shivaani Kummar, James H Doroshow
ABSTRACT
Hypoxia-inducible factor-1 alpha (HIF-1α) is an important marker of hypoxia in human tumors and has been implicated in tumor progression. Drugs targeting HIF-1α are being developed, but the ability to measure drug-induced changes in HIF-1α is limited by the lability of the protein in normoxia. Our goal was to devise methods for specimen collection and processing that preserve HIF-1α in solid tumor tissues and to develop and validate a two-site chemiluminescent quantitative enzyme-linked immunosorbent assay (ELISA) for HIF-1α. We tested various strategies for HIF-1α stabilization in solid tumors, including nitrogen gas-purged lysis buffer, the addition of proteasome inhibitors or the prolyl hydroxylase inhibitor 2-hydroxyglutarate, and bead homogenization. Degassing and the addition of 2-hydroxyglutarate to the collection buffer significantly increased HIF-1α recovery, whereas bead homogenization in sealed tubes improved HIF-1α recovery and reduced sample variability. Validation of the ELISA demonstrated intra- and inter-assay variability of less than 15% and accuracy of 99.8±8.3% as assessed by spike recovery. Inter-laboratory reproducibility was also demonstrated (R(2)=0.999). Careful sample handling techniques allow us to quantitatively detect HIF-1α in samples as small as 2.5μg of total protein extract, and this method is currently being applied to analyze tumor biopsy specimens in early-phase clinical trials.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
L-α-Hydroxyglutaric acid disodium salt, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.