  • Search Results
  • Vitamin D deficiency and its correction in children with sickle cell anaemia.
Vitamin D deficiency and its correction in children with sickle cell anaemia.
Annals of hematology (2014-07-02)
Clare Wykes, Anita Arasaretnam, Sandra O'Driscoll, Laura Farnham, Caje Moniz, David C Rees
ABSTRACT
Vitamin D deficiency is common in sickle cell anaemia (SCA, HbSS), although its significance and optimal means of correction are unknown. We conducted an audit to assess the clinical significance of 25-hydroxy vitamin D (25-OHD) deficiency in children with SCA and to evaluate two methods of vitamin D supplementation. We audited 25-OHD levels in 81 children with SCA and looked for statistical associations with biochemical, haematological and clinical parameters. In a separate group of regularly transfused children with SCA, we compared changes in 25-OHD blood concentrations following treatment with either high-dose intramuscular ergocalciferol (n = 15) or 4 days of high-dose oral cholecalciferol (n = 64). Ninety-one percent of children with SCA had 25-OHD levels <20 μg/L. The 25-OHD levels were negatively correlated with increasing age (P < 0.001) but showed no significant relationship to laboratory measurements, transcranial Doppler velocities or hospital attendance. Both intramuscular ergocalciferol and oral cholecalciferol supplementations resulted in increases of 25-OHD blood concentration to normal levels. The mean dose of ergocalciferol was greater than that of cholecalciferol (7,729 versus 5,234 international units (IU)/kg, P < 0.001), but the increment in 25-OHD levels was significantly greater in the oral cholecalciferol group (6.44 versus 2.82 (ng/L)/(IU/kg), P < 0.001). Both approaches resulted in vitamin D sufficiency for about 120 days. Increased 25-OHD concentration was significantly associated with increased serum calcium concentration. Vitamin D deficiency is very common in SCA and can be effectively corrected with high-dose intramuscular ergocalciferol or 4 days of high-dose oral cholecalciferol. Prospective, randomised studies are needed to assess the clinical value of vitamin D supplementation.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestine, buffered aqueous glycerol solution
Sigma-Aldrich
Phosphatase, Alkaline from bovine kidney, lyophilized powder, ≥5 units/mg protein (Activity determined in diethanolamine buffer, pH 9.8)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥1,000 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, buffered aqueous glycerol solution, ≥5,700 DEA units/mg protein