  • Search Results
  • TGF-beta receptor type-2 expression in cancer-associated fibroblasts regulates breast cancer cell growth and survival and is a prognostic marker in pre-menopausal breast cancer.
TGF-beta receptor type-2 expression in cancer-associated fibroblasts regulates breast cancer cell growth and survival and is a prognostic marker in pre-menopausal breast cancer.
Oncogene (2013-12-18)
S Busch, A Acar, Y Magnusson, P Gregersson, L Rydén, G Landberg
ABSTRACT
Transforming growth factor-beta (TGF-β) is a pleiotropic cytokine with the capability to act as tumour suppressor or tumour promoter depending on the cellular context. TGF-beta receptor type-2 (TGFBR2) is the ligand-binding receptor for all members of the TGF-β family. Data from mouse model experiments demonstrated that loss of Tgfbr2 expression in mammary fibroblasts was linked to tumour initiation and metastasis. Using a randomised tamoxifen trial cohort including in total 564 invasive breast carcinomas, we examined TGFBR2 expression (n=252) and phosphorylation level of downstream target SMAD2 (pSMAD2) (n=319) in cancer-associated fibroblasts (CAFs) and assessed links to clinicopathological markers, prognostic and treatment-predictive values. The study revealed that CAF-specific TGFBR2 expression correlated with improved recurrence-free survival. Multivariate analysis confirmed CAF-TGFBR2 to be an independent prognostic marker (multivariate Cox regression, hazard ratio: 0.534, 95% (CI): 0.360-0.793, P=0.002). CAF-specific pSMAD2 levels, however, did not associate with survival outcome. Experimentally, TGF-β signalling in fibroblasts was modulated using a TGF-β ligand and inhibitor or through lentiviral short hairpin RNA-mediated TGFBR2-specific knockdown. To determine the role of fibroblastic TGF-β pathway on breast cancer cells, we used cell contact-dependent cell growth and clonogenicity assays, which showed that knockdown of TGFBR2 in CAFs resulted in increased cell growth, proliferation and clonogenic survival. Further, in a mouse model transfected CAFs were co-injected with MCF7 and tumour weight and proportion was monitored. We found that mouse xenograft tumours comprising TGFBR2 knockdown fibroblasts were slightly bigger and displayed increased tumour cell capacity. Overall, our data demonstrate that fibroblast-related biomarkers possess clinically relevant information and that fibroblasts confer effects on breast cancer cell growth and survival. Regulation of tumour-stromal cross-talk through fibroblastic TGF-β pathway may depend on fibroblast phenotype, emphasising the importance to characterise tumour microenvironment subtypes.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Trimethylaluminum, 97%