MilliporeSigma
Renin lineage cells repopulate the glomerular mesangium after injury.
Journal of the American Society of Nephrology : JASN (2014-06-07)
Charlotte Starke, Hannah Betz, Linda Hickmann, Peter Lachmann, Björn Neubauer, Jeffrey B Kopp, Maria Luisa S Sequeira-Lopez, R Ariel Gomez, Bernd Hohenstein, Vladimir T Todorov, Christian P M Hugo
ABSTRACT
Mesangial cell injury has a major role in many CKDs. Because renin-positive precursor cells give rise to mesangial cells during nephrogenesis, this study tested the hypothesis that the same phenomenon contributes to glomerular regeneration after murine experimental mesangial injury. Mesangiolysis was induced by administration of an anti-mesangial cell serum in combination with LPS. In enhanced green fluorescent protein-reporter mice with constitutively labeled renin lineage cells, the size of the enhanced green fluorescent protein-positive area in the glomerular tufts increased after mesangial injury. Furthermore, we generated a novel Tet-on inducible triple-transgenic LacZ reporter line that allowed selective labeling of renin cells along renal afferent arterioles of adult mice. Although no intraglomerular LacZ expression was detected in healthy mice, about two-thirds of the glomerular tufts became LacZ positive during the regenerative phase after severe mesangial injury. Intraglomerular renin descendant LacZ-expressing cells colocalized with mesangial cell markers α8-integrin and PDGF receptor-β but not with endothelial, podocyte, or parietal epithelial cell markers. In contrast with LacZ-positive cells in the afferent arterioles, LacZ-positive cells in the glomerular tuft did not express renin. These data demonstrate that extraglomerular renin lineage cells represent a major source of repopulating cells for reconstitution of the intraglomerular mesangium after injury.
Product Description
USP
Enalapril maleate, United States Pharmacopeia (USP) Reference Standard
Enalapril for system suitability, European Pharmacopoeia (EP) Reference Standard
Enalapril maleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Enalapril Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxycycline monohydrate
Sigma-Aldrich
Enalapril maleate salt, powder, ≥98% (TLC)
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
Doxycycline hyclate
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard