  • Search Results
  • Determinants of occupational exposure to metals by gas metal arc welding and risk management measures: a biomonitoring study.
Determinants of occupational exposure to metals by gas metal arc welding and risk management measures: a biomonitoring study.
Toxicology letters (2014-09-17)
Renaud Persoons, Damien Arnoux, Théodora Monssu, Olivier Culié, Gaëlle Roche, Béatrice Duffaud, Denis Chalaye, Anne Maitre
ABSTRACT
Welding fumes contain various toxic metals including chromium (Cr), nickel (Ni) and manganese (Mn). An assessment of the risk to health of local and systemic exposure to welding fumes requires the assessment of both external and internal doses. The aims of this study were to test the relevance in small and medium sized enterprises of a biomonitoring strategy based on urine spot-samples, to characterize the factors influencing the internal doses of metals in gas metal arc welders and to recommend effective risk management measures. 137 welders were recruited and urinary levels of metals were measured by ICP-MS on post-shift samples collected at the end of the working week. Cr, Ni and Mn mean concentrations (respectively 0.43, 1.69 and 0.27 μg/g creatinine) were well below occupational health guidance values, but still higher than background levels observed in the general population, confirming the absorption of metals generated in welding fumes. Both welding parameters (nature of base metal, welding technique) and working conditions (confinement, welding and grinding durations, mechanical ventilation and welding experience) were predictive of occupational exposure. Our results confirm the interest of biomonitoring for assessing health risks and recommending risk management measures for welders.
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Nickel, mesh, 100x250mm, nominal aperture 0.73mm, wire diameter 0.25mm, 26x26 wires/inch, open area 55%, plain weave mesh, 99%
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Nickel, wire reel, 10m, diameter 0.01mm, as drawn, 99.98%