  • Search Results
  • Comparison of bolus injection and constant infusion methods for measuring muscle protein fractional synthesis rate in humans.
Comparison of bolus injection and constant infusion methods for measuring muscle protein fractional synthesis rate in humans.
Metabolism: clinical and experimental (2014-10-14)
Demidmaa Tuvdendorj, David L Chinkes, John Bahadorani, Xiao-jun Zhang, Melinda Sheffield-Moore, Lois A Killewich, Robert R Wolfe
ABSTRACT
The use of stable isotope tracer techniques to measure muscle protein fractional synthesis rate (FSR) has been well established and widely used. The most common method that has been utilized so far is a primed constant infusion (CI) method, which requires 3-4 h of tracer infusion. However, recently our group has developed a bolus injection (BI) method, which requires an injection of bolus of tracer and can be completed within 1 h. In this study, we compared calf (gastrocnemius) muscle protein FSR measured using these two different methods--CI and BI. FSRs were measured in eight people (5 men and 3 women; age: 62.3±6.9 years (mean±SD); body weight: 75.4±21.5 kg) at basal, postabsorptive state using L-[ring-2H5]-phenylalanine. In the CI protocol, a primed continuous infusion was given for 4 h, and muscle biopsies were taken at 120 and 240 min; in the BI, a bolus injection of the tracer was given at 0 min and biopsies were taken at 5 and 60 min. Tracer enrichments in blood and muscle tissue were determined by gas chromatography-mass spectrometry. Data are expressed as mean±SE; t-test, linear regression and Levene Median equal variance test analyses were performed. CI FSR was 0.066±0.006%/h, whereas BI FSR was 0.058±0.008%/h, p=NS. The linear regression analysis showed a significant relationship between BI and CI, p=0.038. The intra-class correlation coefficient was 0.83. The standard deviation of the differences in the measurements was 0.015%/h. The Levene Median equal variance test demonstrated no difference in variance between the CI and BI measurements (p=0.722). No difference could be detected in calf muscle protein FSR measured by CI and BI methods; the BI method can be used for the measurement of muscle protein FSR in humans.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Supelco
L-Phenylalanine, certified reference material, TraceCERT®
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deuterium, 99.9 atom % D
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Phenylalanine, European Pharmacopoeia (EP) Reference Standard