  • Search Results
  • Oxidative stress induces the biosynthesis of citrinin by Penicillium verrucosum at the expense of ochratoxin.
Oxidative stress induces the biosynthesis of citrinin by Penicillium verrucosum at the expense of ochratoxin.
International journal of food microbiology (2014-10-04)
Markus Schmidt-Heydt, Dominic Stoll, Peter Schütz, Rolf Geisen
ABSTRACT
Penicillium verrucosum is a fungus that can produce ochratoxin A and citrinin, two structurally related nephrotoxic mycotoxins. P. verrucosum usually occurs on wheat but can occasionally also be found in NaCl rich habitats such as salted cheeses or olives, indicating that this fungus can adapt to different environments. The ratio of ochratoxin A to citrinin produced by P. verrucosum is shifted to one of either mycotoxin at the expense of the other dependent on the environmental conditions. High NaCl concentrations shift secondary metabolite biosynthesis towards ochratoxin A production. P. verrucosum copes with NaCl stress by increased ochratoxin A biosynthesis, ensuring chloride homeostasis. Ochratoxin A carries chlorine in its molecule and can excrete chlorine from the cell. It was further shown that the regulation of ochratoxin A by high NaCl conditions is mediated by the HOG MAP kinase signal transduction pathway. Here it is shown that high oxidative stress conditions, evoked for example by increasing concentrations of Cu(2+) cations in the growth medium, shift secondary metabolite biosynthesis of P. verrucosum from ochratoxin A to citrinin. The production of citrinin normalizes the oxidative status of the fungal cell under oxidative stress conditions leading to an adaptation to these environmental conditions and protects against increased oxidative stress caused by increased Cu(2+) concentrations. Moreover citrinin also protects against light of short wavelength, which may also increase the oxidative status of the environment. The biosynthesis of citrinin is apparently regulated by a cAMP/PKA signaling pathway, because increasing amounts of external cAMP reduce citrinin biosynthesis in a concentration dependent manner. These conditions lead to the cross-regulation of the ochratoxin A/citrinin secondary metabolite pair and support the adaptation of P. verrucosum to different environments.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Ochratoxin A solution, 10 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Ochratoxin A, from Petromyces albertensis, ≥98% (HPLC)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Ochratoxin A, from Aspergillus ochraceus, reference material
Sigma-Aldrich
Citrinin, from Penicillium citrinum, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Citrinin solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis