  • Search Results
  • Hypernociceptive responses following the intratibial inoculation of RM1 prostate cancer cells in mice.
Hypernociceptive responses following the intratibial inoculation of RM1 prostate cancer cells in mice.
The Prostate (2014-09-30)
María Llorián-Salvador, Marta Pevida, María Teresa Fernández-García, Ana Lastra, Alvaro Obaya, Santiago Cal, Agustín Hidalgo, Luis Menéndez, Ana Baamonde
ABSTRACT
Pain due to bone metastases of prostatic origin is a relevant clinical issue. We study here the nociceptive responses obtained in mice receiving the intratibial inoculation of RM1 prostate cancer cells. 10(2) -10(5) RM1 cells were inoculated to C57BL/6 mice and tumor development was analysed histologically and with luciferase-expressing RM1 cells. Spinal astroglial (GFAP) or microglial (Iba-1) expression was assessed with immunohistochemical methods and hypernociception was measured by the unilateral hot plate, the paw pressure and the von Frey tests. The analgesic effect of morphine, zoledronic acid or the CCR2 antagonist RS504393 was measured. Levels of the chemokines CCL2, CCL3, and CCL5 were determined by ELISA. The inoculation of 10(3) RM1 cells induced tumoral growth in bone with a mixed osteoclastic/osteoblastic pattern and evoked astroglial, but not microglial, activation in the spinal cord. Hyperalgesia and allodynia were already established four days after inoculation and dose-dependently inhibited by the s.c. administration of morphine (1-5 mg/kg) or zoledronic acid (1-3 mg/kg). CCL2 and CCL5, but not CCL3, were released by RM1 cells in culture whereas only an increased presence of CCL2 was found in bone tumor homogenates. The administration of the CCR2 antagonist RS504393 (0.3-3 mg/kg) inhibited RM1 induced thermal hyperalgesia without modifying mechanical allodynia. The intratibial inoculation of RM1 cells in immunocompetent mice induces hypernociceptive responses and can be useful to perform studies of bone cancer induced pain related to androgen-independent prostate cancer. The antinociceptive role derived from the blockade of the CCR2 chemokine receptors is further envisaged.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
SAFC
L-Glutamine
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis