  • Search Results
  • Safety, tolerability, and pharmacokinetics of pregabalin in children with refractory partial seizures: a phase 1, randomized controlled study.
Safety, tolerability, and pharmacokinetics of pregabalin in children with refractory partial seizures: a phase 1, randomized controlled study.
Epilepsia (2014-11-08)
Donald Mann, Jing Liu, Marci L Chew, Howard Bockbrader, Christine W Alvey, Elizabeth Zegarac, John Pellock, Verne W Pitman
ABSTRACT
To evaluate the safety, tolerability, and pharmacokinetics (PK) of pregabalin as adjunctive therapy in children with refractory partial seizures. This was a phase 1, randomized, placebo-controlled, parallel-group, escalating-dose, multiple-dose study comprising a 7-day, double-blind treatment period and a single-blind, single dose of pregabalin administered to all children on day 8. Children in four age cohorts (1-23 months, 2-6, 7-11, and 12-16 years) received one of four doses of pregabalin (2.5, 5, 10, or 15 mg/kg/day) or placebo. Safety and tolerability were assessed throughout the study. Steady-state and single-dose PK parameters on day 8 were analyzed using standard noncompartmental procedures. Sixty-five children received at least one dose of treatment. Four pregabalin-treated children discontinued treatment, three of whom received 15 mg/kg/day. Two children experienced serious adverse events, one of whom received pregabalin 15 mg/kg/day. During double-blind treatment, the most common adverse events reported in the pregabalin-treated population were somnolence (27.1%) and dizziness (12.5%). Steady-state pregabalin peak and total exposure in each age cohort appeared to increase linearly with dose. Apparent oral clearance (CL/F) was directly related to creatinine clearance, consistent with adults. CL/F normalized for body weight was 43% higher in patients weighing <30 kg. Steady-state and single-dose PK were consistent. Pregabalin at doses up to 10 mg/kg/day in children aged 1 month to 16 years, and at doses up to 15 mg/kg/day in those aged <6 years, demonstrated acceptable safety and tolerability. For children weighing <30 kg, a dose increase of 40% (mg/kg dosing) is required to achieve comparable exposure with adults or children weighing ≥30 kg. These data will inform dose selection in phase 3 trials of the efficacy and safety of adjunctive pregabalin in children with refractory partial seizures.
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Supelco
γ-Aminobutyric acid, analytical standard