  • Search Results
  • Intraoperative stereotactic injection of Indigo Carmine dye to mark ill-defined tumor margins: a prospective phase I-II study.
Intraoperative stereotactic injection of Indigo Carmine dye to mark ill-defined tumor margins: a prospective phase I-II study.
Journal of neurosurgery (2014-11-02)
Konstantinos Margetis, Prajwal Rajappa, Apostolos John Tsiouris, Jeffrey P Greenfield, Theodore H Schwartz
ABSTRACT
A critical goal in neurosurgical oncology is maximizing the extent of tumor resection while minimizing the risk to normal white matter tracts. Frameless stereotaxy and white matter mapping are indispensable tools in this effort, but deep tumor margins may not be accurately defined because of the "brain shift" at the end of the operation. The authors investigated the safety and efficacy of a technique for marking the deep margins of intraaxial tumors with stereotactic injection of Indigo Carmine dye. Investigational New Drug study approval for a prospective study in adult patients with gliomas was obtained from the FDA (Investigational New Drug no. 112680). At surgery, 1-3 stereotactic injections of 0.01 ml of Indigo Carmine dye were performed through the initial bur holes into the deep tumor margins before elevation of the bone flap. White light microscopic resection was conducted in standard fashion by using frameless stereotactic navigation until the injected margins were identified. The resection of the injected tumor margins and the extent of resection of the whole tumor volume were determined by using postoperative volumetric MRI. In total 17 injections were performed in 10 enrolled patients (6 male, 4 female), whose mean age was 49 years. For all patients, the injection points were identified intraoperatively and tumor was resected at these points. The staining pattern was reproducible; it was a sphere of stained tissue approximately 5 mm in diameter. A halo of stained tissue and a backflow of dye through the needle tract were also noted, but these were clearly distinct from the staining pattern of the injection point, which was vividly colored and demarcated. Postoperative MR images verified the resection of all injection points. The mean extent of resection of the tumor as a whole was 97.1%. For 1 patient, a brain abscess developed on postoperative Day 16 and needed additional surgical treatment. Stereotactic injection of Indigo Carmine dye can be used to demarcate multiple deep tumor margins, which can be readily identified intraoperatively by using standard white light microscopy. This technique may enhance the accuracy of frameless stereotactic navigation and increase the extent of resection of intraaxial tumors.
Sigma-Aldrich
Indigo, synthetic, Dye content 95 %
Supelco
Indigo carmine, analytical standard
Sigma-Aldrich
Indigo carmine, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Indigo carmine, for microscopy (Bact., Hist.), indicator (pH 11.5-14.0)