  • Search Results
  • Ophiopogonin D attenuates doxorubicin-induced autophagic cell death by relieving mitochondrial damage in vitro and in vivo.
Ophiopogonin D attenuates doxorubicin-induced autophagic cell death by relieving mitochondrial damage in vitro and in vivo.
The Journal of pharmacology and experimental therapeutics (2014-11-08)
Ying-Yu Zhang, Chen Meng, Xin-Mu Zhang, Cai-Hua Yuan, Ming-Da Wen, Zhong Chen, Da-Chuan Dong, Yan-Hong Gao, Chang Liu, Zhao Zhang
ABSTRACT
It has been reported that ophiopogonin D (OP-D), a steroidal glycoside and an active component extracted from Ophiopogon japonicas, promotes antioxidative protection of the cardiovascular system. However, it is unknown whether OP-D exerts protective effects against doxorubicin (DOX)-induced autophagic cardiomyocyte injury. Here, we demonstrate that DOX induced excessive autophagy through the generation of reactive oxygen species (ROS) in H9c2 cells and in mouse hearts, which was indicated by a significant increase in the number of autophagic vacuoles, LC3-II/LC3-I ratio, and upregulation of the expression of GFP-LC3. Pretreatment with OP-D partially attenuated the above phenomena, similar to the effects of treatment with 3-methyladenine. In addition, OP-D treatment significantly relieved the disruption of the mitochondrial membrane potential by antioxidative effects through downregulating the expression of both phosphorylated c-Jun N-terminal kinase and extracellular signal-regulated kinase. The ability of OP-D to reduce the generation of ROS due to mitochondrial damage and, consequently, to inhibit autophagic activity partially accounts for its protective effects in the hearts against DOX-induced toxicity.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Ophiopogonin D, phyproof® Reference Substance
Sigma-Aldrich
Ophiopogonin D, ≥98% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)