  • Search Results
  • The slowdown of the endoglucanase Trichoderma reesei Cel5A-catalyzed cellulose hydrolysis is related to its initial activity.
The slowdown of the endoglucanase Trichoderma reesei Cel5A-catalyzed cellulose hydrolysis is related to its initial activity.
Biochemistry (2014-11-26)
Zhiyu Shu, Yefei Wang, Liaoyuan An, Lishan Yao
ABSTRACT
One important feature of hydrolysis of cellulose by cellulases is that the reaction slows down quickly after it starts. In this work, we investigate the slowdown mechanism at the early stage of the reaction using endoglucanase Tr. Cel5A-catalyzed phosphate acid-swollen cellulose (PASC) hydrolysis as a model system. Specifically, we focus on the effect of enzyme adsorption on the reaction slowdown. Nineteen single mutations are introduced (with the assistance of molecular dynamics simulations) to perturb the enzyme PASC interaction, yielding the adsorption partitioning coefficient Kr that ranged from 0.12 to 0.39 L/g, compared to that of the wild type (0.26 L/g). Several residues, including T18, K26, Y26, H229, and T300, are demonstrated to be important for adsorption of the enzyme to PASC. The kinetic measurements show that the slowdown of the hydrolysis is not correlated with the adsorption quantified by the partitioning coefficient Kr but is anticorrelated with the initial activity. This result suggests that the mutants with higher activity are more prone to being trapped or deplete the most reactive substrate faster and the adsorption plays no apparent role in the reaction slowdown. The initial activity of Cel5A against PASC is correlated with the enzyme specific activity against a soluble substrate p-nitrophenyl cellobioside.
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers