MilliporeSigma
  • Ethanol ablation of predominantly cystic thyroid nodules: evaluation of recurrence rate and factors related to recurrence.
Ethanol ablation of predominantly cystic thyroid nodules: evaluation of recurrence rate and factors related to recurrence.
Clinical radiology (2014-12-03)
C H Suh, J H Baek, E J Ha, Y J Choi, J H Lee, J K Kim, K-W Chung, T Y Kim, W B Kim, Y K Shong
ABSTRACT
To evaluate recurrence rate and associated risk factors for recurrence after ethanol ablation (EA) in patients with predominantly cystic thyroid nodules. This observational study was approved by the Ethics Committee of the Institutional Review Board and informed consent for procedures was obtained. From April 2009 to April 2013, 107 consecutive patients with predominantly cystic nodules were treated using EA. Recurrence was defined as nodules showing a residual solid portion with internal vascularity, cosmetic problems remaining, or persistent symptoms, and patients who requested additional therapy to resolve their symptomatic or cosmetic problems. Delayed recurrence was defined as treated nodules that showed no recurrent features at 1 month, but showed newly developed recurrent features during the longer follow-up period. Multivariate analysis was used for variables to demonstrate the independent factors related to volume reduction. One month after EA, 18.7% of patients (20/107) showed recurrence. Among 87 patients with non-recurrence, 24.1% (21/87) showed delayed recurrence. The total recurrence rate was 38.3% (41/107). Patients with recurrence (n = 41) were treated using radiofrequency ablation (n = 28), second EA (n = 4), and refused further treatment (n = 9). These patients responded well to repeat EA and radiofrequency ablation. Multivariate analysis demonstrated that the initial nodule volume (>20 ml; p < 0.036) and vascularity (grade >1; p < 0.049) were independent predictors of volume reduction at last follow-up. The results revealed that although EA seemed to be effective during the initial period, delayed recurrence should be considered during longer-term follow-up. The independent predictors of recurrence were initial volume (>20 ml) and vascularity.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®