  • Search Results
  • Oncogenic c-Myc and prothymosin-alpha protect hepatocellular carcinoma cells against sorafenib-induced apoptosis.
Oncogenic c-Myc and prothymosin-alpha protect hepatocellular carcinoma cells against sorafenib-induced apoptosis.
Biochemical pharmacology (2014-12-03)
Yi-Te Lin, Hsing-Pang Lu, Chuck C-K Chao
ABSTRACT
Prothymosin alpha (PTMA) is overexpressed in various human tumors, including hepatocellular carcinoma (HCC). The significance of PTMA overexpression and its underlying mechanism remain unclear. We show here that silencing PTMA sensitizes HCC cells to the kinase inhibitor sorafenib. In contrast, ectopic expression of PTMA induces cell resistance to the drug. While inhibitors targeting JNK, ERK or PI3K reduce PTMA expression, only ERK activation is suppressed by sorafenib. In addition, inhibition of ERK produces a dramatic decrease in both endogenous PTMA level and promoter activation. Ectopic expression of active MKK1/2 considerably induces PTMA expression. We also identify a sorafenib-responsive segment lying 1000-1500-bp upstream of the PTMA transcription start site and observe that it is controlled by c-Myc and ERK. Mutation in the PTMA promoter at the predicted c-Myc binding site and silencing of c-Myc both abrogate sorafenib's effect on PTMA transcription. We also find that silencing PTMA potentiates Bax translocation to mitochondria in response to sorafenib and this is associated with increased cytochrome c release from mitochondria and enhanced caspase-9 activation. These results indicate that PTMA is positively regulated by the oncoprotein c-Myc and protects HCC cells against sorafenib-induced cell death, thus identifying PTMA as a new target for chemotherapy against HCC.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
2-Butanone, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
2-Butanone, analytical standard
SAFC
Sodium deoxycholate
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
2-Butanone, ReagentPlus®, ≥99%
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®