  • Search Results
  • MiR-22 regulates 5-FU sensitivity by inhibiting autophagy and promoting apoptosis in colorectal cancer cells.
MiR-22 regulates 5-FU sensitivity by inhibiting autophagy and promoting apoptosis in colorectal cancer cells.
Cancer letters (2014-12-03)
Honghe Zhang, Jinlong Tang, Chen Li, Jianlu Kong, Jingyu Wang, Yihua Wu, Enping Xu, Maode Lai
ABSTRACT
Autophagy has become one of the most important mechanisms of chemotherapy resistance by supporting the survival of tumor cells under metabolic and therapeutic stress. Here, we showed that miR-22 inhibited autophagy and promoted apoptosis to increase the sensitivity of colorectal cancer (CRC) cells to 5-fluorouracil (5-FU) treatment both in vitro and in vivo. B-cell translocation gene 1 (BTG1) was identified as a new target of miR-22, which could reverse the inhibition of autophagy induced by miR-22. Thus, miR-22 may function as an important switch between autophagy and apoptosis to regulate 5-FU sensitivity through post-transcriptional silencing of BTG1. Promisingly, miR-22 could be considered as both a predictor of 5-FU sensitivity for personalized treatment and a therapeutic target for colorectal cancer.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Supelco
5-Fluorouracil, analytical standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorouracil, meets USP testing specifications