  • Search Results
  • Optimization of factors affecting hexavalent chromium removal from simulated electroplating wastewater by synthesized magnetite nanoparticles.
Optimization of factors affecting hexavalent chromium removal from simulated electroplating wastewater by synthesized magnetite nanoparticles.
Environmental monitoring and assessment (2014-12-05)
Mitra Ataabadi, Mehran Hoodaji, Arezoo Tahmourespour, Mahmoud Kalbasi, Majid Abdouss
ABSTRACT
Hexavalent chromium is a mutagen and carcinogen that is of significant concern in water and wastewater. In the present study, magnetite nanoparticles (n-Mag) were investigated as a potential remediation technology for the decontamination of Cr (VI)-contaminated wastewater. Synthesized n-Mag was characterized by X-ray diffraction (XRD), scanning electron microscopy (SEM), and BET-N2 technology. To screen and optimize the factors affecting Cr (VI) removal efficiency by synthesized nanoparticles, Plackett-Burman (PB) and Taguchi experimental designs were used respectively. The crystalline produced n-Mag was in the size range of 60-70 nm and had a specific surface area (SSA) of 31.55 m(2) g(-1). Results of PB design showed that the most significant factors affecting Cr (VI) removal efficiency were initial Cr (VI) concentration, pH, n-Mag dosage, and temperature. In a pH of 2, 20 mg L(-1) of Cr (VI) concentration, 4 g L(-1)of n-Mag, temperature of 40 °C, 220 rpm of shaking speed, and 60 min of contact time, the complete removal efficiency of Cr (VI) was achieved. Batch experiments revealed that the removal of Cr (VI) by n-Mag was consistent with pseudo-second order reaction kinetics. The competition from common coexisting ions such as NO₃(-), SO₄(2-), and Cl(-) were not considerable, unless in the higher concentration of SO₄(2-). These results indicated that the readily synthesized magnetite nanoparticles have promising applications for the removal of Cr (VI) from aqueous solution.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Chromium, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 99.99+%