  • Search Results
  • Association of common SIX6 polymorphisms with peripapillary retinal nerve fiber layer thickness: the Singapore Chinese Eye Study.
Association of common SIX6 polymorphisms with peripapillary retinal nerve fiber layer thickness: the Singapore Chinese Eye Study.
Investigative ophthalmology & visual science (2014-12-30)
Ching-Yu Cheng, R Rand Allingham, Tin Aung, Yih-Chung Tham, Michael A Hauser, Eranga N Vithana, Chiea Chuen Khor, Tien Yin Wong
ABSTRACT
Recently the common SIX6 missense variant rs33912345 was found to be highly associated with glaucoma. The aim of this study was to investigate the association between this SIX6 variant and peripapillary retinal nerve fiber layer (RNFL) thickness measured by spectral-domain optical coherence tomography (SD-OCT) in a population setting. Study subjects were enrolled from the Singapore Chinese Eye Study (SCES), a population-based survey of Singaporean Chinese aged 40 years or older. Subjects underwent a comprehensive ocular examination. Spectral-domain OCT was used to measure RNFL thicknesses. Genotyping of SIX6 rs33912345 (Asn141His) was performed using HumanExome BeadChip. A total of 2129 eyes from 1243 SCES subjects (mean age: 55.0 ± 7.4 years) with rs33912345 genotype data and SD-OCT images were included for the analysis. Of these, 26 eyes of 21 subjects had glaucoma. The frequency of rs33912345 risk variant C (His141) was 80% in the study subjects. Each rs33912345 C allele was associated with a decrease of 1.44 μm in RNFL thickness after adjusting for age, sex, genetic principal components, and axial length (P = 0.001). These associations remained similar in 2096 nonglaucoma eyes in which each C allele was associated with a decrease of 1.39 μm in RNFL thickness (P = 0.001). The strongest association was observed in the superior RNFL sector (a decrease of 2.83 μm per risk allele, P < 0.001) followed by the inferior RNFL sector (a decrease of 2.24 μm per risk allele, P = 0.003), while the association did not reach significance in the nasal and temporal sectors. Nonglaucomatous individuals with the SIX6 missense variant have reduced RNFL thickness in regions known to be particularly affected in those with glaucoma. This may be the primary mechanism for increased risk of POAG in individuals who carry the SIX6 His141 risk variant.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization