  • Search Results
  • Flapless alveolar ridge preservation utilizing the "socket-plug" technique: clinical technique and review of the literature.
Flapless alveolar ridge preservation utilizing the "socket-plug" technique: clinical technique and review of the literature.
The Journal of oral implantology (2014-12-17)
Georgios Kotsakis, Vanessa Chrepa, Nicolas Marcou, Hari Prasad, James Hinrichs
ABSTRACT
It has been documented that after every extraction of one or more teeth, the alveolar bone of the respective region undergoes resorption and atrophy. Therefore, ridge preservation techniques are often employed after tooth extraction to limit this phenomenon. The benefits of a flapless procedure include maintenance of the buccal keratinized gingiva, prevention of alterations to the gingival contours, and migration of the mucogingival junction that are often experienced after raising a flap. The purpose of this article is to review the literature concerning flapless ridge preservation techniques with the aid of collagen plugs for occlusion of the socket. The term "socket-plug" technique is introduced to describe these techniques. The basic steps of the "socket-plug" technique consist of atraumatic tooth extraction, placement of the appropriate biomaterials in the extraction site, preservation of soft tissue architecture employing a flapless technique, and placement and stabilization of the collagen plug. A case example is presented that illustrates the steps used in this technique.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium silicate, purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Calcium silicate, −200 mesh, 99%
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.