MilliporeSigma
Anti-septic effects of fisetin in vitro and in vivo.
Inflammation (2014-04-08)
Hayoung Yoo, Sae-Kwang Ku, Min-Su Han, Kyung-Min Kim, Jong-Sup Bae
ABSTRACT
Sepsis is a state of disrupted inflammatory homeostasis that is initiated by infection. High mobility group box 1 (HMGB1) protein acting as a late mediator of severe vascular inflammatory conditions, such as sepsis and endothelial cell protein C receptor (EPCR), is involved in vascular inflammation. Fisetin, an active compound from the family Fabaceae, was reported to have antiviral, neuroprotective, and anti-inflammatory activities. Here, we determined the anti-septic effects of fisetin on HMGB1-mediated inflammatory responses and on the shedding of EPCR in vitro and in vivo, for the first time. First, we monitored the effects of post-treatment fisetin on lipopolysaccharide (LPS) and cecal ligation and puncture (CLP)-mediated release of HMGB1 and HMGB1-mediated regulation of pro-inflammatory responses in human umbilical vein endothelial cells (HUVECs) and septic mice. Post-treatment fisetin was found to suppress LPS-mediated release of HMGB1 and HMGB1-mediated cytoskeletal rearrangements. Fisetin also inhibited HMGB1-mediated hyperpermeability and leukocyte migration in septic mice. Fisetin induced potent inhibition of phorbol-12-myristate 13-acetate (PMA) and CLP-induced EPCR. Fisetin also inhibited the expression and activity of tumor necrosis factor-α converting enzyme, induced by PMA in endothelial cells. In addition, fisetin inhibited the production of tumor necrosis factor-α and the activation of AKT, nuclear factor-κB, and extracellular regulated kinases 1/2 by HMGB1 in HUVECs. Fisetin also down-regulated CLP-induced release of HMGB1, production of interleukin 1β, and reduced septic mortality. Collectively, these results suggest that fisetin may be a candidate therapeutic agent for the treatment of vascular inflammatory diseases via inhibition of the HMGB1 signaling pathway.
Sigma-Aldrich
HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-RELA (Ab-468) antibody produced in rabbit, affinity isolated antibody
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mouse KC / CXCL1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human GRO-α / CXCL1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human Procalcitonin ELISA Kit
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Rat CINC-1 ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Anti-RELA (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-276) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-435) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-311) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-536) antibody produced in rabbit, affinity isolated antibody