MilliporeSigma
  • Roflumilast N-oxide inhibits bronchial epithelial to mesenchymal transition induced by cigarette smoke in smokers with COPD.
Roflumilast N-oxide inhibits bronchial epithelial to mesenchymal transition induced by cigarette smoke in smokers with COPD.
Pulmonary pharmacology & therapeutics (2014-02-15)
Javier Milara, Teresa Peiró, Adela Serrano, Ricardo Guijarro, Cristóbal Zaragozá, Herman Tenor, Julio Cortijo
ABSTRACT
Epithelial to mesenchymal transition (EMT) is under discussion as a potential mechanism of small airway remodelling in COPD. In bronchial epithelium of COPD and smokers markers of EMT were described. In vitro, EMT may be reproduced by exposing well-differentiated human bronchial epithelial cells (WD-HBEC) to cigarette smoke extract (CSE). EMT may be mitigated by an increase in cellular cAMP. This study explored the effects of roflumilast N-oxide, a PDE4 inhibitor on CSE-induced EMT in WD-HBEC and in primary bronchial epithelial cells from smokers and COPD in vitro. WD-HBEC from normal donors were stimulated with CSE (2.5%) for 72 h in presence of roflumilast N-oxide (2 nM or 1 μM) or vehicle. mRNA and protein of EMT markers αSMA, vimentin, collagen-1, E-cadherin, ZO-1, KRT5 as well as NOX4 were quantified by real-time quantitative PCR or protein array, respectively. Phosphorylated and total ERK1/2 and Smad3 were assessed by protein array. cAMP and TGFβ1 were measured by ELISA. Reactive oxygen species (ROS) were determined by DCF fluorescence, after 30 min CSE (2.5%). Apoptosis was measured with Annexin V/PI labelling. In some experiments, EMT markers were determined in monolayers of bronchial epithelial cells from smokers, COPD versus controls. Roflumilast N-oxide protected from CSE-induced EMT in WD-HBEC. The PDE4 inhibitor reversed both the increase in mesenchymal and the loss in epithelial EMT markers. Roflumilast N-oxide restored the loss in cellular cAMP following CSE, reduced ROS, NOX4 expression, the increase in TGFβ1 release, phospho ERK1/2 and Smad3. The PDE4 inhibitor partly protected from the increment in apoptosis with CSE. Finally the PDE4 inhibitor decreased mesenchymal yet increased epithelial phenotype markers in HBEC of COPD and smokers. Roflumilast N-oxide may mitigate epithelial-mesenchymal transition in bronchial epithelial cells in vitro.
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Roflumilast, ≥98% (HPLC)
Sigma-Aldrich
PhosphoDetect Anti-Smad3 (pSer423/425) Rabbit mAb (C25A9), liquid, clone C25A9, Calbiochem®
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Vimentin antibody, Mouse monoclonal, clone V9, purified from hybridoma cell culture