  • Search Results
  • Exendin-4 inhibits HMGB1-induced inflammatory responses in HUVECs and in murine polymicrobial sepsis.
Exendin-4 inhibits HMGB1-induced inflammatory responses in HUVECs and in murine polymicrobial sepsis.
Inflammation (2014-05-16)
Wonhwa Lee, Sae-Kwang Ku, Eun Ji Park, Dong Hee Na, Kyung-Min Kim, Jong-Sup Bae
ABSTRACT
Exendin-4 (EX4) has been reported to attenuate myocardial ischemia and reperfusion (I/R) injury and inflammatory and oxidative responses. Nuclear DNA-binding protein high-mobility group box 1 (HMGB1) protein acts as a late mediator of severe vascular inflammatory conditions. However, the effect of EX4 on HMGB1-induced inflammatory response has not been studied. First, we accessed this question by monitoring the effects of posttreatment EX4 on lipopolysaccharide (LPS) and cecal ligation and puncture (CLP)-mediated release of HMGB1 and HMGB1-mediated regulation of proinflammatory responses in human umbilical vein endothelial cells (HUVECs) and septic mice. Posttreatment EX4 was found to suppress LPS-mediated release of HMGB1 and inhibited HMGB1-mediated hyperpermeability and leukocyte migration in septic mice. EX4 also induced downregulation of CLP-induced release of HMGB1, production of IL-6, and mortality. Collectively, these results suggest that EX4 may be regarded as a candidate therapeutic agent for treatment of vascular inflammatory diseases via inhibition of the HMGB1 signaling pathway.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Anti-RELA (Ab-311) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-536) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-435) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-276) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-505) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Anti-RELA (Ab-468) antibody produced in rabbit, affinity isolated antibody
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine