  • Search Results
  • 2-Deoxy-d-glucose attenuates sevoflurane-induced neuroinflammation through nuclear factor-kappa B pathway in vitro.
2-Deoxy-d-glucose attenuates sevoflurane-induced neuroinflammation through nuclear factor-kappa B pathway in vitro.
Toxicology in vitro : an international journal published in association with BIBRA (2014-06-08)
Qingxiu Wang, Yupeng Zhao, Min Sun, Sheng Liu, Baolin Li, Lei Zhang, Longqiu Yang
ABSTRACT
Sevoflurane, one of the most commonly used anesthetics in clinic, induced neuroinflammation and caused cognitive impairment. 2-deoxy-d-glucose (2-DG) is a synthetic analogue of glucose and is clinically used in medical imaging safely. We examined the effect of 2-DG on sevoflurane-induced neuroinflammation in the mouse primary microglia cells. Mouse microglia cells were treated with 4.1% sevoflurane for 6h to examine the expression of interleukin (IL)-6 and tumor necrosis factor (TNF-α) and activation of nuclear factor-kappa B (NF-κB). Pyrrolidine dithiocarbamate (PDTC) or 2-DG was used 1h before sevoflurane treatment. In the present study, we found that sevoflurane increased level of IL-6 and TNF-α through activating NF-κB signaling, and that 2-DG reduced sevoflurane-induced increase in IL-6 and TNF-α and nuclear NF-κB in microglia cells. Our data suggests that NF-κB signaling pathway could be a target for sevoflurane-induced neuroinflammation and 2-DG might be a potential therapy to prevent or treat sevoflurane-induced neuroinflammation.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sevoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant