  • Search Results
  • Involvement of extracellular factors in maintaining self-renewal of neural stem cell by nestin.
Involvement of extracellular factors in maintaining self-renewal of neural stem cell by nestin.
Neuroreport (2014-06-12)
Chun Guang Di, Andy Peng Xiang, Lei Jia, Jun Feng Liu, Bruce T Lahn, Bao Feng Ma
ABSTRACT
Nestin knockout leads to embryonic lethality and self-renewal deficiency in neural stem cells (NSCs). However, how nestin maintains self-renewal remains uncertain. Here, we used the dosage effect of nestin in heterozygous mice (Nes+/-) to study self-renewal of NSCs. With existing extracellular signaling in vivo or in vitro, nestin levels do not affect proliferation ability or apoptosis when compared between Nes+/- and Nes+/+ NSCs. However, self-renewal ability of Nes+/- NSCs is impaired when plated at a low cell density and completely lost at a clonal density. This deficiency in self-renewal at a clonal density is rescued using a medium conditioned by Nes+/+ NSCs. In addition, the Akt signaling pathway is altered at low density and reversed by conditioned medium. Our data show that secreted factors contribute toward maintaining self-renewal of NSCs by nestin, potentially through Akt signaling.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
DAPI, dilactate, ≥98% (HPLC)
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Rabbit anti-AKT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.