  • Search Results
  • Trim28 contributes to EMT via regulation of E-cadherin and N-cadherin in lung cancer cell lines.
Trim28 contributes to EMT via regulation of E-cadherin and N-cadherin in lung cancer cell lines.
PloS one (2014-07-02)
Lu Chen, Teresita Muñoz-Antonia, W Douglas Cress
ABSTRACT
In previous work, we demonstrated that transcription factor Trim28 (Tripartite motif containing 28) plays a tumor-suppressor role in early-staged adenocarcinoma of the lung due to its ability to restrain transcription of cell cycle-regulating genes. Herein we examine Trim28's role in the epithelial-to-mesenchymal transition (EMT) which is strongly implicated in cancer metastasis. We found that Trim28 plays a role in TGF-β-induced EMT in non-small cell lung cancer cells. Silencing Trim28 with inhibitory RNAs alters the expression of numerous EMT markers, such as E-cadherin and N-cadherin, whereas overexpression of Trim28 has an opposite effect. Trim28 expression is induced following TGF-β treatment at both protein and mRNA levels. Trim28 deficiency impairs TGF-β-induced EMT and decreases cell migration and invasion. Finally, we demonstrate that the expression of Trim28 affects the acetylation and methylation of histones on E-cadherin and N-cadherin promoters. These results suggest that Trim28 contributes to EMT and might be important for tumor metastasis in lung cancer. Taken together with our previous work these results suggest a model in which Trim28 is a tumor suppressor early in the transformation process in lung cancer, but in later stages it functions as an oncogene.
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Cdh1 antibody,Mouse monoclonal, clone DCS-266, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, serum, Upstate®
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution