  • Search Results
  • Rrd1p, an RNA polymerase II-specific prolyl isomerase and activator of phosphoprotein phosphatase, promotes transcription independently of rapamycin response.
Rrd1p, an RNA polymerase II-specific prolyl isomerase and activator of phosphoprotein phosphatase, promotes transcription independently of rapamycin response.
Nucleic acids research (2014-08-13)
Rwik Sen, Shivani Malik, Sarah Frankland-Searby, Bhawana Uprety, Shweta Lahudkar, Sukesh R Bhaumik
ABSTRACT
Rrd1p (resistance to rapamycin deletion 1) has been previously implicated in controlling transcription of rapamycin-regulated genes in response to rapamycin treatment. Intriguingly, we show here that Rrd1p associates with the coding sequence of a galactose-inducible and rapamycin non-responsive GAL1 gene, and promotes the association of RNA polymerase II with GAL1 in the absence of rapamycin treatment following transcriptional induction. Consistently, nucleosomal disassembly at GAL1 is impaired in the absence of Rrd1p, and GAL1 transcription is reduced in the Δrrd1 strain. Likewise, Rrd1p associates with the coding sequences of other rapamycin non-responsive and inducible GAL genes to promote their transcription in the absence of rapamycin treatment. Similarly, inducible, but rapamycin-responsive, non-GAL genes such as CTT1, STL1 and CUP1 are also regulated by Rrd1p. However, transcription of these inducible GAL and non-GAL genes is not altered in the absence of Rrd1p when the steady-state is reached after long transcriptional induction. Consistently, transcription of the constitutively active genes is not changed in the Δrrd1 strain. Taken together, our results demonstrate a new function of Rrd1p in stimulation of initial rounds of transcription, but not steady-state/constitutive transcription, of both rapamycin-responsive and non-responsive genes independently of rapamycin treatment.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Methylbutanol, puriss. p.a., ACS reagent, ≥98.5% (GC)
Supelco
3-Methyl-1-butanol, analytical standard
Supelco
3-Methylbutanol, analytical standard