  • Search Results
  • Periostin secreted by epithelial ovarian carcinoma is a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins and promotes cell motility.
Periostin secreted by epithelial ovarian carcinoma is a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins and promotes cell motility.
Cancer research (2002-09-18)
Lindsay Gillan, Daniela Matei, David A Fishman, C S Gerbin, Beth Y Karlan, David D Chang
ABSTRACT
Periostin (PN) is a secreted protein that shares a structural homology to the axon guidance protein fasciclin I in insects. Previously, we reported that PN expression is up-regulated in epithelial ovarian tumors. We further examined the role of PN in ovarian cancer. PN is expressed in several normal tissues but not in normal ovaries and has a tendency for higher expression in fetal tissues. Ovarian cancer cells secrete PN, which can accumulate in malignant ascites of ovarian cancer patients. Purified recombinant PN supports adhesion of ovarian epithelial cells that can be inhibited by monoclonal antibodies against alpha(V)beta(3) or alpha(V)beta(5) integrin, but not by anti-beta(1) integrin antibody. Furthermore, alpha(V)beta(3) integrin, but not beta(1) integrins, colocalizes to the focal adhesion plaques formed on PN. Cells plated on PN form fewer stress fibers and are more motile compared with those plated on fibronectin. We propose PN functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins to support adhesion and migration of ovarian epithelial cells.
Sigma-Aldrich
Anti-POSTN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.