  • Search Results
  • Periostin, a member of a novel family of vitamin K-dependent proteins, is expressed by mesenchymal stromal cells.
Periostin, a member of a novel family of vitamin K-dependent proteins, is expressed by mesenchymal stromal cells.
The Journal of biological chemistry (2008-05-03)
Daniel L Coutu, Jian Hui Wu, Anne Monette, Georges-Etienne Rivard, Mark D Blostein, Jacques Galipeau
ABSTRACT
The modification of glutamic acid residues to gamma-carboxyglutamic acid (Gla) is a post-translational modification catalyzed by the vitamin K-dependent enzyme gamma-glutamylcarboxylase. Despite ubiquitous expression of the gamma-carboxylation machinery in mammalian tissues, only 12 Gla-containing proteins have so far been identified in humans. Because bone tissue is the second most abundant source of Gla-containing proteins after the liver, we sought to identify Gla proteins secreted by bone marrow-derived mesenchymal stromal cells (MSCs). We used a proteomics approach to screen the secretome of MSCs with a combination of two-dimensional gel electrophoresis and tandem mass spectrometry. The most abundant Gla-containing protein secreted by MSCs was identified as periostin, a previously unrecognized gamma-carboxylated protein. In silico amino acid sequence analysis of periostin demonstrated the presence of four consensus gamma-carboxylase recognition sites embedded within fasciclin-like protein domains. The carboxylation of periostin was confirmed by immunoprecipitation and purification of the recombinant protein. Carboxylation of periostin could be inhibited by warfarin in MSCs, demonstrating its dependence on the presence of vitamin K. We were able to demonstrate localization of carboxylated periostin to bone nodules formed by MSCs in vitro, suggesting a role in extracellular matrix mineralization. Our data also show that another fasciclin I-like protein, betaig-h3, contains Gla. In conclusion, periostin is a member of a novel vitamin K-dependent gamma-carboxylated protein family characterized by the presence of fasciclin domains. Furthermore, carboxylated periostin is produced by bone-derived cells of mesenchymal lineage and is abundantly found in mineralized bone nodules in vitro.
Sigma-Aldrich
Anti-POSTN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.