  • Search Results
  • MafB: an activator of the glucagon gene expressed in developing islet alpha- and beta-cells.
MafB: an activator of the glucagon gene expressed in developing islet alpha- and beta-cells.
Diabetes (2006-01-31)
Isabella Artner, John Le Lay, Yan Hang, Lynda Elghazi, Jonathan C Schisler, Eva Henderson, Beatriz Sosa-Pineda, Roland Stein
ABSTRACT
The large Maf family of basic leucine-zipper-containing transcription factors are known regulators of key developmental and functional processes in various cell types, including pancreatic islets. Here, we demonstrate that within the adult pancreas, MafB is only expressed in islet alpha-cells and contributes to cell type-specific expression of the glucagon gene through activation of a conserved control element found between nucleotides -77 to -51. MafB was also shown to be expressed in developing alpha- and beta-cells as well as in proliferating hormone-negative cells during pancreatogenesis. In addition, MafB expression is maintained in the insulin(+) and glucagon(+) cells remaining in mice lacking either the Pax4 or Pax6 developmental regulators, implicating a potentially early role for MafB in gene regulation during islet cell development. These results indicate that MafB is not only important to islet alpha-cell function but may also be involved in regulating genes required in both endocrine alpha- and beta-cell differentiation.
Sigma-Aldrich
Anti-MAFB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.