  • Search Results
  • Photoswitchable fluorescent proteins for superresolution fluorescence microscopy circumventing the diffraction limit of light.
Photoswitchable fluorescent proteins for superresolution fluorescence microscopy circumventing the diffraction limit of light.
Methods in molecular biology (Clifton, N.J.) (2013-10-11)
Susana Rocha, Herlinde De Keersmaecker, Hiroshi Uji-i, Johan Hofkens, Hideaki Mizuno
ABSTRACT
In the last two decades, fluorescent proteins became an indispensable tool to noninvasively label a protein in living cells. The discovery of photoswitchable fluorescent proteins expanded the applications of the fluorescent proteins to techniques such as molecular tracking and highlighting on a microscope. Recently, a new microscopic modality to achieve a superresolution circumventing the diffraction limit of light (photoactivated localization microscopy, PALM) has been developed based on the photoswitchable fluorescent proteins. Here we introduce a basic protocol of PALM through the visualization of actin bundles with superresolution.
Product Description
Roche
X-tremeGENE 9 DNA Transfection Reagent, Polymer reagent for transfecting common cell lines
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.