  • Search Results
  • Infrared intensities of liquids. XVII. Infrared refractive indices from 8000 to 350 cm-1, absolute integrated absorption intensities, transition moments, and dipole moment derivatives of methan-d3-ol and methanol-d4 at 25?C.
Infrared intensities of liquids. XVII. Infrared refractive indices from 8000 to 350 cm<SUP>-1</SUP>, absolute integrated absorption intensities, transition moments, and dipole moment derivatives of methan-<I>d</I><SUB>3</SUB>-ol and methanol-<I>d</I><SUB>4</SUB> at 25?C.
J. Chem. Phys. (1994)
Bertie JE and Zhang SL.