  • Search Results
  • Intranasal deferoxamine engages multiple pathways to decrease memory loss in the APP/PS1 model of amyloid accumulation.
Intranasal deferoxamine engages multiple pathways to decrease memory loss in the APP/PS1 model of amyloid accumulation.
Neuroscience letters (2014-12-03)
Jared M Fine, Daniel B Renner, Anna C Forsberg, Rachel A Cameron, Benjamin T Galick, Clint Le, Patrick M Conway, Benjamin M Stroebel, William H Frey, Leah R Hanson
ABSTRACT
In addition to the hallmark accumulation of amyloid and hyper-phosphorylation of tau, brain changes in Alzheimer's disease are multifactorial including inflammation, oxidative stress, and metal dysregulation. Metal chelators have been explored as a less well known approach to treatment. One chelator currently being developed is deferoxamine (DFO), administered via the intranasal (IN) route. In the current study, APP/PS1 amyloid mice were treated with a chronic, low dose of IN DFO, subjected to a rigorous battery of behavior tests, and the mechanism of action was examined. Mice were treated 3x/week with 0.24 C IN DFO for 18 weeks from 36 to 54 weeks of age, 4 weeks of behavior tests were performed that included both working and reference memory, anxiolytic and motor behaviors, and finally brain tissues were analyzed for amyloid, protein oxidation, and other proteins affected by DFO. We found that IN DFO treatment significantly decreased loss of both reference and working memory in the Morris and radial arm water mazes (p < 0.05), and also decreased soluble Aβ40 and Aβ42 in cortex and hippocampus (p < 0.05). Further, IN DFO decreased activity of GSK3β, and led to decreases in oxidative stress (p < 0.05). These data demonstrate that low doses of IN DFO can modify several targets along the multiple pathways implicated in the neuropathology of Alzheimer's, making it an attractive candidate for the treatment of this heterogeneous disease.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
SAFC
Sodium deoxycholate
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)