  • Search Results
  • Seminal parameters of chronic male genital inflammation are associated with disturbed sperm DNA integrity.
Seminal parameters of chronic male genital inflammation are associated with disturbed sperm DNA integrity.
Andrologia (2015-02-25)
F Haidl, G Haidl, I Oltermann, J P Allam
ABSTRACT
Definition of chronic male genital tract inflammation and its impact on male infertility is still a matter of debate. In particular, DNA integrity has been reported to be disturbed in subfertile men. Thus, the aim of this study was to investigate an association of DNA integrity to altered standard semen parameters as well as inflammatory parameters such as peroxidase-positive cells, macrophages and seminal interleukin-6 concentration. Macrophages were detected by CD18/HLA-Dr staining, and DNA integrity was analysed by acridine orange staining using flow cytometry. Interleukin-6 was detected by ELISA. Normal DNA integrity showed a significant correlation to sperm number and progressive motility. Moreover, a significant inverse correlation of DNA integrity to Interleukin-6 and macrophages could be demonstrated. Further on, seminal interleukin-6 also significantly correlated to macrophages. No association has been observed between the number of peroxidase-positive cells and normal DNA integrity. As disturbed DNA integrity has been reported to negatively influence spermatozoon-egg interaction and even fertilisation rates following ICSI, and as early miscarriages have been associated with sperm DNA damage, it should be screened very carefully for male genital tract inflammations in couples undergoing infertility treatment. Measuring Interleukin-6 seems superior to assessment of the number of leucocytes alone and additional assessment of DNA integrity into the diagnostic work-up should be considered.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)