  • Search Results
  • Influence of Brucella abortus lipopolysaccharide as an adjuvant on the immunogenicity of HPV-16 L1VLP vaccine in mice.
Influence of Brucella abortus lipopolysaccharide as an adjuvant on the immunogenicity of HPV-16 L1VLP vaccine in mice.
Medical microbiology and immunology (2014-09-05)
Zahra Kianmehr, Hoorieh Soleimanjahi, Susan Kaboudanian Ardestani, Fatemeh Fotouhi, Asghar Abdoli
ABSTRACT
Brucella abortus lipopolysaccharide (LPS) has less toxicity and no pyrogenic properties in comparison with other bacterial LPS. It is a toll-like receptor 4 agonist and has been shown to have the potential use as a vaccine adjuvant. In this study, the immunostimulatory properties of LPS from smooth and rough strains of B. abortus (S19 and RB51) as adjuvants were investigated for the human papillomavirus type 16 (HPV16) L1 virus-like particles (L1VLPs) vaccines. C57BL/6 mice were immunized subcutaneously three times either with HPV-16 L1VLPs alone, or in combination with smooth LPS (S-LPS), rough LPS (R-LPS), aluminum hydroxide or a mixture of them as adjuvant. The humoral immunity was evaluated by measuring the specific and total IgG levels, and also the T-cell immune response of mice was evaluated by measuring different cytokines such as IFN-γ, TNF-α, IL-4, IL-10 and IL-17. Results showed that serum anti-HPV16 L1VLP IgG antibody titers was significantly higher in mice immunized with a combination of VLPs and R-LPS or S-LPS compared with other immunized groups. Co-administration of HPV-16 L1VLPs with R-LPS elicited the highest levels of splenocytes cytokines (IFN-γ, IL-4, IL-17 and TNF-α) and also effectively induced improvement of a Th1-type cytokine response characterized with a high ratio of IFN-γ/IL-10. The data indicate that B. abortus LPS particularly RB51-LPS enhances the immune responses to HPV-16 L1VLPs and suggests its potential as an adjuvant for the development of a potent prophylactic HPV vaccine and other candidate vaccines.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC