  • Search Results
  • Immunomodulatory effects of galectin-1 on an IgE-mediated allergic conjunctivitis model.
Immunomodulatory effects of galectin-1 on an IgE-mediated allergic conjunctivitis model.
Investigative ophthalmology & visual science (2015-01-13)
Claudia Bosnic Mello, Lisandra Ramos, Alexandre Dantas Gimenes, Teresa Raquel de Moraes Andrade, Sonia Maria Oliani, Cristiane Damas Gil
ABSTRACT
Galectin (Gal)-1, a lectin found at sites of immune privilege with critical role in the inflammation, has been poorly investigated in the ocular inflammatory diseases. Here, we evaluated the therapeutic potential of Gal-1 in ocular allergy using a model of ovalbumin (OVA)-induced AC. OVA-immunized BALB/c male mice were challenged with eye drops containing OVA on days 14 through 16 with a subset of animals pretreated intraperitoneally with recombinant Gal-1 (rGal-1) or dexamethasone (Dex). Recombinant Gal-1 and Dex administration on days 14 through 16 was effective in reducing the clinical signs of allergic conjunctivitis (AC), plasma anti-OVA IgE levels, Th2 (IL-4 and IL-13), and eotaxin/RANTES levels in the lymph nodes. Four hours after the last OVA challenge, rGal-1 markedly increased Gal-1 endogenous levels in the conjunctiva, and provoked eosinophilia, which persisted at 24 hours. Recombinant Gal-1 had no effect on eosinophil activation, as evidenced by the similar pattern of peroxidase eosinophil expression between cells of rGal-1-treated and untreated AC groups. Conjunctival migrated eosinophils and neutrophils exhibited high levels of Gal-1 and β2-integrin, with points of colocalization, in the rGal-1-treated groups. These different effects observed for rGal-1 were correlated with elevated levels of activated ERK and p38 at 4 hours, and diminished levels of activated JNK and p38 at 24 hours in the eyes. Gal-1 has an important role in ocular allergic inflammation and represents a potential target for the development of new therapeutic strategies in eye diseases.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
SAFC
Sodium chloride solution, 5 M
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 32 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%