  • Search Results
  • Protective effects of Iranian Achillea wilhelmsii essential oil on acetaminophen-induced oxidative stress in rat liver.
Protective effects of Iranian Achillea wilhelmsii essential oil on acetaminophen-induced oxidative stress in rat liver.
Pharmaceutical biology (2014-09-23)
A Dadkhah, F Fatemi, M Alipour, Z Ghaderi, F Zolfaghari, F Razdan
ABSTRACT
Achillea wilhelmsii C. Koch (Asteraceae) is widely used in Iranian traditional medicine. This in vivo study evaluates the hepatoprotective role of Iranian A. wilhelmsii oils against acetaminophen-induced oxidative damages in rats. The animals were divided into five groups: in negative control and control groups, the DMSO and 500 mg/kg acetaminophen were i.p. injected, respectively. In treatment groups, 100 and 200 mg/kg oils and 10 mg/kg BHT were given i.p. immediately after acetaminophen administration. Then, the hepatic oxidative/antioxidant parameters such as lipid peroxidation (LP), glutathione (GSH), superoxide dismutase (SOD), catalase (CAT), and ferric reducing ability of plasma (FRAP) were measured in time intervals (2, 4, 8, 16, and 24 h) after administrations confirmed by histophatological consideration at 24 h. The results indicated that acetaminophen caused a significant elevation in SOD activity (8-24 h) and LP and FRAP levels (4 h) paralleled with significant decline in GSH level (4 and 8 h). The apparent oxidative injury was associated with evident hepatic necrosis confirmed in histological examination. The presences of A. wilhelmsii oils (100 and 200 mg/kg) with acetaminophen mitigated significantly the rise in SOD, LP, and FRAP levels and restored the GSH compared with the group treated with acetaminophen. These were confirmed by histological examination indicating the hepatic necrosis reversal by the oils. It can be concluded that concomitant administration of A. wilhelmsii oils with acetaminophen may be useful in reversing the drug hepatotoxicity.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Glutathione reduced, BioReagent, suitable for cell culture, ≥98.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%