  • Search Results
  • Osteopontin, osteocalcin and OB-cadherin expression in Synthetic nanohydroxyapatite vs bovine hydroxyapatite cultured Osteoblastic-like cells.
Osteopontin, osteocalcin and OB-cadherin expression in Synthetic nanohydroxyapatite vs bovine hydroxyapatite cultured Osteoblastic-like cells.
Journal of biological regulators and homeostatic agents (2014-10-16)
A Santarelli, M Mascitti, G Orsini, L Memè, R Rocchetti, P Tiriduzzi, F Sampalmieri, A Putignano, M Procaccini, L Lo Muzio, F Bambini
ABSTRACT
Calcium phosphate ceramics have been applied in bone replacement for several decades due to their excellent biocompatibility, bioactivity, osteo-conductivity and mechanical strength. Several studies have demonstrated that porous hydroxyapatite (HA) is an excellent scaffold for osteogenic proliferation and differentiation of the osteoprogenitor cells. However, different methods of synthesis and production of HA ceramic-based materials may have considerable effect on the physical and biological properties. In the present work, two hydroxyapatite-based materials, a natural hydroxyapatite ceramic of bovine origin and a synthetic nano-cristalline hydroxyapatite were tested in vitro with MG63 cell line. The results displayed that both the materials demonstrated a good biocompatibility. The immunocytochemical stain revealed a different positivity of the osteogenic markers between the cultures with the biomaterials, and the control culture. Western blot data confirmed the immunocytochemical stain. Both the materials tested in the present study demonstrated a good biocompatibility with the osteoblastic cells allowing, at the same time, the osteogenic differentiation, and they may be useful in clinical use.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
2-Propanol, analytical standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade