  • Search Results
  • Fragile TIM-4-expressing tissue resident macrophages are migratory and immunoregulatory.
Fragile TIM-4-expressing tissue resident macrophages are migratory and immunoregulatory.
The Journal of clinical investigation (2014-07-02)
Thomas B Thornley, Zemin Fang, Savithri Balasubramanian, Rafael A Larocca, Weihua Gong, Shipra Gupta, Eva Csizmadia, Nicolas Degauque, Beom Seok Kim, Maria Koulmanda, Vijay K Kuchroo, Terry B Strom
ABSTRACT
Macrophages characterized as M2 and M2-like regulate immune responses associated with immune suppression and healing; however, the relationship of this macrophage subset to CD169+ tissue-resident macrophages and their contribution to shaping alloimmune responses is unknown. Here we identified a population of M2-like tissue-resident macrophages that express high levels of the phosphatidylserine receptor TIM-4 and CD169 (TIM-4hiCD169+). Labeling and tracking of TIM-4hiCD169+ macrophages in mice revealed that this population is a major subset of tissue-resident macrophages, homes to draining LNs following oxidative stress, exhibits an immunoregulatory and hypostimulatory phenotype that is maintained after migration to secondary lymphoid organs, favors preferential induction of antigen-stimulated Tregs, and is highly susceptible to apoptosis. Moreover, CD169+ tissue-resident macrophages were resistant to oxidative stress-induced apoptosis in mice lacking TIM-4. Compared with heart allografts from WT mice, Tim4-/- heart allografts survived much longer and were more easily tolerized by non-immunosuppressed recipients. Furthermore, Tim4-/- allograft survival was associated with the infiltration of Tregs into the graft. Together, our data provide evidence that M2-like TIM-4hiCD169+ tissue-resident macrophages are immunoregulatory and promote engraftment of cardiac allografts, but their influence is diminished by TIM-4-dependent programmed cell death.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
DBP, 98% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Monoclonal Anti-IL2RA antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Monoclonal Anti-CD25 antibody produced in mouse, clone MEM-140, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD25 antibody produced in mouse, clone MEM-181, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, Selectophore
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MONOCLONAL ANTI-CD25 antibody produced in mouse, clone 7G7B6, crude ascites, buffered aqueous solution