  • Search Results
  • Synergistic cell growth inhibition by the combination of amrubicin and Akt-suppressing agents in K-ras mutation-harboring lung adenocarcinoma cells: implication of EGFR tyrosine kinase inhibitors.
Synergistic cell growth inhibition by the combination of amrubicin and Akt-suppressing agents in K-ras mutation-harboring lung adenocarcinoma cells: implication of EGFR tyrosine kinase inhibitors.
International journal of oncology (2014-01-09)
Shizuka Ito, Tadashi Igishi, Miyako Takata, Yasuto Ueda, Shingo Matsumoto, Masahiro Kodani, Kenichi Takeda, Hiroki Izumi, Tomohiro Sakamoto, Kosuke Yamaguchi, Haruhiko Makino, Hirokazu Touge, Hiroki Chikumi, Eiji Shimizu
ABSTRACT
Previously we showed that Akt-suppressing agents, combined with amrubicin, synergistically inhibited the growth of small cell lung cancer cells. The combined effects of chemotherapeutic agents and Akt-suppressing agents, including epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors, were evaluated in A549 lung adenocarcinoma cells harboring K-ras mutation and wild-type EGFR. Only amrubicin and not other chemotherapeutics (cisplatin, pemetrexed and paclitaxel) synergistically inhibited cell growth when combined with an Akt inhibitor, LY294002. The combination of amrubicin and LY294002 enhanced Annexin V binding to cells. A non-specific tyrosine kinase inhibitor, genistein, suppressed Akt and showed synergistic interaction in combination with amrubicin. Two EGFR tyrosine kinase inhibitors (EGFR-TKIs), gefitinib and erlotinib, suppressed Akt activity at clinically achievable concentrations and demonstrated synergism when combined with amrubicin. The suppression of K-ras expression by siRNA interfered with this synergism and inhibited both EGFR and Akt activity in A549 cells. In Ma10 cells, which harbor wild-type EGFR and K-ras, EGFR-TKIs neither suppressed Akt activity nor exhibited such synergism when combined with amrubicin. We concluded that the synergism by the combination of EGFR-TKI and amrubicin is attributable, at least partially, to K-ras mutation in A549 cells. The combination of EGFR-TKI and amrubicin may be a promising treatment for lung cancer with wild-type EGFR and K-ras mutation.
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody