MilliporeSigma
Reproductive toxicity of nanoscale graphene oxide in male mice.
Nanotoxicology (2014-03-14)
Shanlu Liang, Shun Xu, Ding Zhang, Junmin He, Maoquan Chu
ABSTRACT
In the past few years, much work has been performed to explore the biomedical applications and toxicity of nano-graphene and its derivatives. However, the reproductive toxicity of those carbon nanomaterials has been rarely studied. In this study, we report on the male reproductive toxicity of nanoscale graphene oxide (GO) using a mouse model. The results showed that the adult male mice injected with high dosages of GO (25 mg/kg mouse) via the tail vein exhibited normal sex hormone secretion and retained normal reproductive activity. All untreated female mice mated with the GO-treated male mice could produce healthy pups. There were no significant differences in pup numbers, sex ratio, weights, pup survival rates or pup growth over time between the GO-treated and control groups. Furthermore, these GO-treated male mice could produce a second, third, fourth and even fifth litter of healthy offspring when they lived with the untreated female mice. The testicular and epididymal histology as well as the activities of several important epididymal enzymes including α-glucosidase, lactate dehydrogenase, glutathione peroxidase and acid phosphatase were not affected by GO treatment. In addition, no damaging effects were seen at high dose rates of GO (total 300 mg/kg male mouse, 60 mg/kg every 24 h for 5 days) via intra-abdominal injection. Thus, GO showed very low or nearly no toxicity for male reproduction. This work will greatly enable future investigations of GO nanosheets for in vivo biomedical applications.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Potassium permanganate, puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)