  • Search Results
  • A stability-indicating HPLC method for the quantification of aliskiren and hydrochlorothiazide in a pharmaceutical formulation.
A stability-indicating HPLC method for the quantification of aliskiren and hydrochlorothiazide in a pharmaceutical formulation.
Journal of AOAC International (2015-01-30)
Dimitrios Karvelis, Eleni Kalogria, Irene Panderi
ABSTRACT
A novel, fast, and sensitive stability-indicating HPLC method was developed, fully validated, and applied to the simultaneous determination of aliskiren and hydrochlorothiazide in a combined formulation. Effective chromatographic separation was achieved using a phenyl analytical column with isocratic elution using the mobile phase 0.030 M ammonium acetate-acetonitrile (60 + 40, v/v) at a flow rate of 0.40 mL/min. The UV spectrophotometric detector was set at 280 nm. The method was linear over the concentration ranges of 1.5-4.5 and 0.125-0.375 μg/mL for aliskiren and hydrochlorothiazide, respectively. The intraday and interday RSD values were less than 6.1%, while the relative percentage error, Er, was less than 5% for both analytes. Both drugs were subjected to stress conditions of acidic and alkaline hydrolysis, oxidation, and thermal degradation. The proposed method proved to be stability indicating by resolution of the drugs from their forced degradation products. The method was applied successfully to the QC and content uniformity tests in combined commercial tablets.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.