  • Search Results
  • The role of matrix metalloproteinases and neutrophil gelatinase-associated lipocalin in central and peripheral arterial aneurysms.
The role of matrix metalloproteinases and neutrophil gelatinase-associated lipocalin in central and peripheral arterial aneurysms.
Surgery (2014-12-03)
Raffaele Serra, Raffaele Grande, Rossella Montemurro, Lucia Butrico, Francesco Giuseppe Caliò, Diego Mastrangelo, Edoardo Scarcello, Luca Gallelli, Gianluca Buffone, Stefano de Franciscis
ABSTRACT
An association between arterial aneurysms and matrix metalloproteinases (MMPs) has been described previously. MMPs regulate extracellular structural proteins and tissue remodeling. Neutrophil gelatinase-associated lipocalin (NGAL) is involved in the regulation of MMP activity. The aim of this work was to study the relationship between the levels of MMPs and NGAL and arterial aneurysms. In a multicenter, open-label, parallel groups, prospective study, patients with aneurysmal disease were divided into 2 groups: Group I (with ruptured aneurysm) and group II (with nonruptured aneurysm). Healthy volunteer patients were also enrolled and represented the control group (group III). We enrolled 307 patients (107 in group I and 200 in group II) with arterial aneurysm: 49 popliteal, 31 common femoral, 2 superficial femoral, 29 common iliac artery, 3 common carotid, and 193 abdominal aorta. Finally, 11 healthy volunteer patients (9 males and 2 females; age range, 40-70 years; median 56) were enrolled in group III. Enzyme-linked immunosorbent assay and Western blot analysis revealed greater levels of immunoreactive MMP-9 and NGAL in all patients with ruptured aneurysms, both central and peripheral aneurysms, and in the aneurismal vessels. These results provide potentially important insights to the understanding of the natural history of arterial aneurysms. MMPs and NGAL play a role in development of arterial aneurysms and may represent molecular markers for the prevention of aneurysmal rupture.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
BICINE, ≥99% (titration)
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.