MilliporeSigma
Modern diagnostic approach to hereditary xanthinuria.
Urolithiasis (2014-11-06)
Martin Mraz, Olha Hurba, Josef Bartl, Zdenek Dolezel, Anthony Marinaki, Lynette Fairbanks, Blanka Stiburkova
ABSTRACT
Hereditary xanthinuria (HX) is a rare inherited disorder caused by a deficiency of xanthine dehydrogenase/oxidase (XDH/XO). Missing XDH/XO activity leads to undetectable levels of uric acid excessively replaced by xanthine in serum/urine. The allopurinol loading test has been traditionally used to differentiate between HX types I and II. Final confirmation of HX has been based on the biopsy finding of the absent XDH/XO activity in the small intestine or liver. We present the clinical, biochemical, ultrasound and molecular genetics findings in three new patients with HX and suggest a simple three-step approach to be used for diagnosis, typing and confirmation of HX. In the first step, the diagnosis of HX is determined by extremely low serum/urinary uric acid excessively replaced by xanthine. Second, HX is typed using urinary metabolomics. Finally, the results are confirmed by molecular genetics. We advocate for this safe and non-invasive diagnostic algorithm instead of the traditional allopurinol loading test and intestinal or liver biopsy used in the past.
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis