  • Search Results
  • KAP-1 is overexpressed and correlates with increased metastatic ability and tumorigenicity in pancreatic cancer.
KAP-1 is overexpressed and correlates with increased metastatic ability and tumorigenicity in pancreatic cancer.
Medical oncology (Northwood, London, England) (2014-05-28)
Chao Yu, Lei Zhan, Jianxin Jiang, Yaozhen Pan, Hong Zhang, Xu Li, Feng Pen, Min Wang, Renyi Qin, Chenyi Sun
ABSTRACT
This study aimed to investigate the role in metastasis and prognostic value of KAP-1 in pancreatic cancer (PC). The expression of KAP-1 was analyzed by quantitative real-time polymerase chain reaction, Western blotting, and immunohistochemical staining in 91 human PC tissue samples. Capan-2 cells were transfected with a lentiviral vector expressing KAP-1 (Capan-2/KAP-1) or the empty vector (Capan-2/vector); cell migration and invasion were assayed in vitro using Transwell migration and wound-healing assays, and in vivo using a xenograft model in nude mice. KAP-1 was found to be overexpressed in human PC, and the expression of KAP-1 correlated with clinical stage. Overexpression of KAP-1 increased the invasion and migration of Capan-2 cells in vitro. Furthermore, overexpression of KAP-1 promoted the growth and metastatic ability of PC cells in a xenograft model in nude mice. Moreover, overexpression of KAP-1 induced the epithelial-mesenchymal transition (EMT) in PC cells both in vitro and in vivo, as indicated by increased expression of mesenchymal markers such as vimentin and decreased expression of E-cadherin. This study indicates that KAP-1 may promote metastasis in PC by regulating the EMT and suggests that KAP-1 may have potential as a predictor of metastasis in patients with pancreatic cancer.
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CDH2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Cdh1 antibody,Mouse monoclonal, clone DCS-266, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CDH2 antibody produced in mouse, clone 5C8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-N-Cadherin antibody produced in mouse, clone GC-4, ascites fluid
Sigma-Aldrich
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution