  • Search Results
  • Dual immune modulatory effect of vitamin A in human visceral leishmaniasis.
Dual immune modulatory effect of vitamin A in human visceral leishmaniasis.
PloS one (2014-10-01)
Bruna L Lima Maciel, Bruna L Lima Maciel, Joanna Gardel Valverde, João Firmino Rodrigues-Neto, Francisco Freire-Neto, Tatjana S L Keesen, Selma Maria Bezerra Jeronimo
ABSTRACT
Vitamin A supplementation has shown to prevent mortality by diarrheal and respiratory diseases in several countries. Nevertheless, there are few studies investigating the effect of vitamin A in visceral leishmaniasis (VL), although there are reports of its deficiency in children with symptomatic VL in Brazil and Bangladesh. This study analyzed the effect of vitamin A on a subset of Treg cells and monocytes isolated from symptomatic VL and from healthy children residing in an endemic area for VL in Northeast Brazil. Serum retinol concentrations correlated inversely with IL-10 and TGF-β productions in CD4(+)CD25(high)Foxp3(+) T cells isolated from children with VL stimulated with leishmanial antigens. All-trans retinoic acid in vitro induced IL-10 in CD4(+)CD25(high)Foxp3(+) T cells; IL-10 and TGF-β production in CD4(+)CD25-Foxp3- T cells, and IL-10 in monocytes isolated from healthy children. However, the use of all-trans retinoic acid together with leishmanial antigens in vitro prevented increases in IL-10 production in Treg cells and monocytes isolated from VL children. Strikingly, those results show a potential dual role of vitamin A in the immune system: improvement of a regulatory profile in cells from healthy children after leishmanial stimulation and down modulation of IL-10 in Treg cells and monocytes during symptomatic VL. Therefore, the use of vitamin A concomitant to VL therapy might be useful in improving recovery from disease status caused by Leishmania infantum infection and warrants additional study.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Hexane, analytical standard