  • Search Results
  • HIV-1 induces B-cell activation and class switch recombination via spleen tyrosine kinase and c-Jun N-terminal kinase pathways.
HIV-1 induces B-cell activation and class switch recombination via spleen tyrosine kinase and c-Jun N-terminal kinase pathways.
AIDS (London, England) (2014-08-28)
Ana Judith Perisé-Barrios, Rafael Correa-Rocha, Susana Alvarez, Maria Angeles Muñoz-Fernandez, Marjorie Pion
ABSTRACT
Patients infected by the HIV type 1 (HIV-1) frequently show a general deregulation of immune system. A direct influence of HIV-1 particles on B-cell activation, proliferation and B-cell phenotype alterations has been recently described. Moreover, expression of activation-induced cytidinedeaminase (AID) mRNA, which is responsible for class switch recombination (CSR) and somatic hypermutation (SHM), was reported to be overexpressed in B cells exposed to HIV-1. Study of primary human B cells in an in-vitro model. In the current study, we evaluated which signalling pathways are activated in primary B cells after a direct contact with HIV-1 particles in vitro using different kinase inhibitors. Here, we report that B-cell activation together with the increase of AID mRNA expression and the subsequent class switch recombination (CSR) in HIV-exposed B cells occurred through spleen tyrosine kinase (SYK) and c-Jun N-terminal kinase (JNK) pathways. Therefore, we showed that HIV-1 could directly induce primary B-cell deregulation via SYK/B-cell receptor (BCR) engagement, and that activation was followed by the JNK pathway activation. To our knowledge, these data provide the first evidence that SYK/BCR activation was the first step for B-cell activation and CSR mechanism after HIV-1 stimulation in a T-cell-free context.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Supelco
Sucrose, analytical standard
Sucrose, European Pharmacopoeia (EP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin, analytical standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%