  • Search Results
  • Rosmarinic acid exerts an antiosteoporotic effect in the RANKL-induced mouse model of bone loss by promotion of osteoblastic differentiation and inhibition of osteoclastic differentiation.
Rosmarinic acid exerts an antiosteoporotic effect in the RANKL-induced mouse model of bone loss by promotion of osteoblastic differentiation and inhibition of osteoclastic differentiation.
Molecular nutrition & food research (2014-11-08)
Ji-Won Lee, Midori Asai, Sang-Kyung Jeon, Tadahiro Iimura, Takayuki Yonezawa, Byung-Yoon Cha, Je-Tae Woo, Akira Yamaguchi
ABSTRACT
Bone homeostasis is ensured by the balance between bone formation and resorption. Thus, control of the recruitment, proliferation, and differentiation of bone cells is essential to maintain bone mass. The aim of this study was to elucidate the effects of rosmarinic acid as a potential therapeutic agent on bone metabolism using bone cells and a mouse model. Rosmarinic acid increased alkaline phosphatase activity and induced mineralization in osteoblasts. Addition of rosmarinic acid to cultures of calvarial osteoblastic cells prepared from T-cell factor/β-catenin TOP-GAL mutant mice strongly induced the expression of LacZ and promoted stabilization of β-catenin in the cytoplasm of ST2 cells, suggesting that rosmarinic acid affects the canonical Wnt signaling pathway. Moreover, rosmarinic acid inhibited not only osteoclast formation in cocultures of mouse bone marrow cells and osteoblasts, but also receptor activator of nuclear factor kappa-B ligand (RANKL)-induced osteoclastic differentiation in bone marrow-derived macrophages. RANKL-induced p38 mitogen-activated protein kinase and the expression of nuclear factor of activated T cell, c-Jun, and c-Fos were inhibited by rosmarinic acid in bone marrow macrophages. Finally, we confirmed that rosmarinic acid improved bone mass in a soluble RANKL-induced bone loss mouse model. Rosmarinic acid has dual regulatory effects on bone metabolism and may control the bone functions by controlling osteoblastic and osteoclastic differentiation.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, reagent grade