IL-17 promotes murine lupus.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-13)
Gil Amarilyo, Elaine V Lourenço, Fu-Dong Shi, Antonio La Cava
ABSTRACT
The proinflammatory activity of IL-17-producing Th17 cells has been associated with the pathogenesis of several autoimmune diseases. In this article, we provide direct evidence for a role of IL-17 in the pathogenesis of systemic lupus erythematosus (SLE). The induction of SLE by pristane in IL-17-sufficient wild-type mice did not occur in IL-17-deficient mice, which were protected from development of lupus autoantibodies and glomerulonephritis. The protection from SLE in IL-17-deficient mice was associated with a reduced frequency of CD3(+)CD4(-)CD8(-) double-negative T cells and an expansion of CD4(+) regulatory T cells, and did not depend on Stat-1 signaling. These data affirm the key role of IL-17 in the pathogenesis of SLE and strengthen the support for IL-17 blockade in the therapy of SLE.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Pristane, synthetic, ≥98% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
SAFC
Pristane, synthetic, liquid, sterile-filtered, BioReagent
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC