  • Search Results
  • PTPN22 controls virally-induced autoimmune diabetes by modulating cytotoxic T lymphocyte responses in an epitope-specific manner.
PTPN22 controls virally-induced autoimmune diabetes by modulating cytotoxic T lymphocyte responses in an epitope-specific manner.
Clinical immunology (Orlando, Fla.) (2014-12-17)
Georgia Fousteri, Tatiana Jofra, Roberta Di Fonte, Mirela Kuka, Matteo Iannacone, Manuela Battaglia
ABSTRACT
Ptpn22 is one of the most potent autoimmunity predisposing genes and strongly associates with type 1 diabetes (T1D). Previous studies showed that non-obese diabetic mice with reduced expression levels of Ptpn22 are protected from T1D due to increased number of T regulatory (Treg) cells. We report that lack of Ptpn22 exacerbates virally-induced T1D in female rat insulin promoter lymphocytic choriomeningitis virus (RIP-LCMV-GP) mice, while maintaining higher number of Treg cells throughout the antiviral response in the blood and spleen but not in the pancreatic lymph nodes. GP33-41-specific pentamer-positive cytotoxic lymphocytes (CTLs) are numerically reduced in the absence of Ptpn22 at the expansion and contraction phase but reach wild-type levels at the memory phase. However, they show similar effector function and even a subtle increase in the production of IL-2. In contrast, NP396-404-specific CTLs develop normally at all phases but display enhanced effector function. Lack of Ptpn22 also augments the memory proinflammatory response of GP61-80 CD4 T cells. Hence, lack of Ptpn22 largely augments antiviral effector T cell responses, suggesting that caution should be taken when targeting Ptpn22 to treat autoimmune diseases where viral infections are considered environmental triggers.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC