  • Search Results
  • LL-37 peptide enhancement of signal transduction by Toll-like receptor 3 is regulated by pH: identification of a peptide antagonist of LL-37.
LL-37 peptide enhancement of signal transduction by Toll-like receptor 3 is regulated by pH: identification of a peptide antagonist of LL-37.
The Journal of biological chemistry (2014-08-06)
Divyendu Singh, Robert Vaughan, C Cheng Kao
ABSTRACT
LL-37 is a peptide secreted by human epithelial cells that can lyse bacteria, suppress signaling by Toll-like receptor 4 (TLR4), and enhance signaling to double-stranded RNA (dsRNA) by TLR3. How LL-37 interacts with dsRNA to affect signal transduction by TLR3 is not completely understood. We determined that LL-37 binds dsRNA and traffics to endosomes and releases the dsRNA in a pH-dependent manner. Using dynamic light scattering spectroscopy and cell-based FRET experiments, LL-37 was found to form higher order complexes independent of dsRNA binding. Upon acidification LL-37 will dissociate from a larger complex. In cells, LL-37 has a half-live of ∼ 1 h. LL-37 half-life was increased by inhibiting endosome acidification or inhibiting cathepsins, which include proteases whose activity are activated by endosome acidification. Residues in LL-37 that contact poly(I:C) and facilitate oligomerization in vitro were mapped. Peptide LL-29, which contains the oligomerization region of LL-37, inhibited LL-37 enhancement of TLR3 signal transduction. LL-29 prevented LL-37 · poly(I:C) co-localization to endosomes containing TLR3. These results shed light on the requirements for LL-37 enhancement of TLR3 signaling.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)