  • Search Results
  • C1QBP negatively regulates the activation of oncoprotein YBX1 in the renal cell carcinoma as revealed by interactomics analysis.
C1QBP negatively regulates the activation of oncoprotein YBX1 in the renal cell carcinoma as revealed by interactomics analysis.
Journal of proteome research (2014-12-17)
Yong Wang, Dan Yue, Mingming Xiao, Can Qi, Yajing Chen, Duxin Sun, Ning Zhang, Ruibing Chen
ABSTRACT
The Y-box-binding protein 1 (YBX1) plays a critical role in tumorigenesis by promoting cell proliferation, overriding cell-cycle check points, and enhancing genomic instability. In this study, the interactome of YBX1 in renal cell carcinoma (RCC) was analyzed by coimmunoprecipitation and mass spectrometry to better understand its function and regulatory mechanism. A total of 129 proteins were identified as potential YBX1 binding partners. The interaction between the complement component 1, q subcomponent binding protein (C1QBP), and YBX1 was further confirmed by immunoprecipitation and Western blotting. Knockdown of C1QBP enhanced the phosphorylation of YBX1and its nuclear translocation, indicating that C1QBP negatively regulated YBX1 activation. The clinical significance of these two proteins was analyzed in the tissues from 52 RCC patients by immunohistochemistry. Expression of YBX1 was markedly elevated in the carcinoma tissues, and its nuclear expression was associated with histological T stage and metastasis. Meanwhile, the level of C1QBP in the carcinoma tissues was significantly lower than that in the adjacent healthy tissues, which was negatively correlated with the nuclear localization of YBX1 in the RCC tissues (P = 0.011). These data suggest that C1QBP is a novel regulator of YBX1, and the expression of C1QBP and the nuclear expression of YBX1 could both be used as independent prognostic makers for cancer progression in the RCC patients. The proteomics data have been deposited to the ProteomeXchange with identifier PXD001493.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
SAFC
Iodoacetamide
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%